missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromic Acid, Fisher Chemical
Click to view available options
Quantity:
1 L
Packaging:
Plastic Coated Glass bottle
Specifications
Specifications
pH | 0.3 |
Color | Colorless to Yellow |
Melting Point | 10°C |
Boiling Point | 290°C |
Packaging | Plastic Coated Glass bottle |
Quantity | 1 L |
Physical Form | Liquid |
MDL Number | 10952 |
Safety and Handling
Product Identifier
- Chromic Acid
- Danger
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Carcinogenicity Category 1B
- LONG-TERM AQUATIC HAZARD Chronic 3
- H314-Causes severe skin burns and eye damage.
- H350i-May cause cancer by inhalation.
- H412-Harmful to aquatic life with long lasting effects.
- EUH208-Contains A§. May produce an allergic reaction.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/A§.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction