missing translation for 'onlineSavingsMsg'
Learn More
Chromium, 99+%, Thermo Scientific Chemicals Product Code.: 15219726

Chromium, 99+%, Thermo Scientific Chemicals

Product Code. 15219726
100 g
Click to view available options
Quantity:
100 g
500 g
2.5 kg
This item is not returnable. View return policy

Product Code. 15219726

Brand: Thermo Scientific Acros 225491000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-47-3
Molecular Formula Cr
Molecular Weight (g/mol) 52.00
MDL Number MFCD00010944
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
Synonym chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
PubChem CID 23976
ChEBI CHEBI:28073
IUPAC Name chromium
SMILES [Cr]

Specifications

Melting Point 1857.0°C
Boiling Point 2642.0°C
Density 7.2000g/mL
Linear Formula Cr
Quantity 100 g
Solubility Information Solubility in water: insoluble.
Formula Weight 52
Percent Purity ≥99%
Grade Pure
Chemical Name or Material Chromium

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463545 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number225491000,225495000
Lot NumberA0458222 Certificate TypeCertificate of Analysis Date06/12/2023 Catalog Number225491000,225490025,225495000
Lot NumberA0452590 Certificate TypeCertificate of Analysis Date05/05/2023 Catalog Number225491000,225495000
Lot NumberA0396760 Certificate TypeCertificate of Analysis Date16/12/2022 Catalog Number225491000,225490025,225495000
Lot NumberA0413865 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number225491000,225490025,225495000
compliance-icons
Product Identifier
  • Chromium
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
Hazard Statement
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P391-Collect spillage.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chromium, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.