Learn More
Chromium carbide, 99.5% (metals basis), Thermo Scientific Chemicals
59.80 € - 230.15 €
Chemical Identifiers
CAS | 12012-35-0 |
---|---|
Molecular Formula | C2Cr3 |
Molecular Weight (g/mol) | 180.01 |
MDL Number | MFCD00016041 |
InChI Key | UFGZSIPAQKLCGR-UHFFFAOYSA-N |
Synonym | chromium carbide, chromic carbide, chromium carbide,-325 mesh, amperit? 580.002, chromium 2+ ;methanidylidynechromium |
PubChem CID | 3650773 |
IUPAC Name | chromium(2+);methanidylidynechromium |
SMILES | [Cr]#C[Cr]C#[Cr] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11307257
|
Thermo Scientific Alfa Aesar
011547.18 |
50 g |
59.80 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11397247
|
Thermo Scientific Alfa Aesar
011547.30 |
250 g |
230.15 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chromium carbide is useful in the surface treatment of metal components. It is used to coat the surface of another metal in a technique known as thermal spraying. It is also used as an additive in cutting tools made out of cemented carbides, in order to improve toughness by preventing the growth of large grains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12012-35-0 | |
180.01 | |
UFGZSIPAQKLCGR-UHFFFAOYSA-N | |
3650773 | |
[Cr]#C[Cr]C#[Cr] |
C2Cr3 | |
MFCD00016041 | |
chromium carbide, chromic carbide, chromium carbide,-325 mesh, amperit? 580.002, chromium 2+ ;methanidylidynechromium | |
chromium(2+);methanidylidynechromium |
Specifications
1890°C | |
Powder | |
MFCD00016041 | |
chromium carbide, chromic carbide, chromium carbide,-325 mesh, amperit? 580.002, chromium 2+ ;methanidylidynechromium | |
UFGZSIPAQKLCGR-UHFFFAOYSA-N | |
chromium(2+);methanidylidynechromium | |
3650773 | |
99.5% | |
(metals basis) |
12012-35-0 | |
C2Cr3 | |
UN3178 | |
Insoluble in water. | |
[Cr]#C[Cr]C#[Cr] | |
180.01 | |
180.02 | |
Odorless | |
Chromium carbide |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 234-576-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.