missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium carbide, 99.5% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Chromium carbide is useful in the surface treatment of metal components. Chromium carbide is used to coat the surface of another metal in a technique known as thermal spraying. Chromium carbide is used as an additive in cutting tools made out of cemented carbides, in order to improve toughness by preventing the growth of large grains.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Gray |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 99.5% (Metal basis) |
Solubility Information | Reacts with water. |
Formula Weight | 1267.97 |
Odor | Odorless |
Appearance | Gray powder |
Chemical Name or Material | Chromium carbide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'