Learn More
Chromium hexacarbonyl, 99+%, Thermo Scientific Chemicals
132.00 € - 451.00 €
Chemical Identifiers
CAS | 13007-92-6 |
---|---|
Molecular Formula | C6CrO6 |
Molecular Weight (g/mol) | 220.06 |
MDL Number | MFCD00010945 |
InChI Key | KOTQLLUQLXWWDK-UHFFFAOYSA-N |
Synonym | chromium hexacarbonyl, chromium carbonyl, chromium 0 hexacarbonyl, carbon monoxide; chromium, hexacarbonyl chromium, unii-920f3ejx2p, ccris 3186, chromium carbonyl oc-6-11, hexacarbonylchromium 10g |
PubChem CID | 25589 |
ChEBI | CHEBI:33031 |
IUPAC Name | carbon monoxide;chromium |
SMILES | [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10687362
|
Thermo Scientific Acros
221050100 |
10 g | Glass bottle |
132.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10542744
|
Thermo Scientific Acros
221050500 |
50 g | Glass bottle |
451.00 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13007-92-6 | |
220.06 | |
KOTQLLUQLXWWDK-UHFFFAOYSA-N | |
25589 | |
carbon monoxide;chromium |
C6CrO6 | |
MFCD00010945 | |
chromium hexacarbonyl, chromium carbonyl, chromium 0 hexacarbonyl, carbon monoxide; chromium, hexacarbonyl chromium, unii-920f3ejx2p, ccris 3186, chromium carbonyl oc-6-11, hexacarbonylchromium 10g | |
CHEBI:33031 | |
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr] |
Specifications
220°C | |
13007-92-6 | |
Crystalline Powder and/or Crystals | |
C6CrO6 | |
MFCD00010945 | |
chromium hexacarbonyl, chromium carbonyl, chromium 0 hexacarbonyl, carbon monoxide; chromium, hexacarbonyl chromium, unii-920f3ejx2p, ccris 3186, chromium carbonyl oc-6-11, hexacarbonylchromium 10g | |
KOTQLLUQLXWWDK-UHFFFAOYSA-N | |
carbon monoxide;chromium | |
25589 | |
220.06 | |
Authentic |
150°C | |
White through Yellow | |
99+% | |
Cr(CO)6 | |
15, 2236 | |
Solubility in water: insoluble. Other solubilities: sparingly soluble in organic solvents | |
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Cr] | |
220.06 | |
CHEBI:33031 | |
≥99% | |
Chromium hexacarbonyl |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
Harmful if inhaled.
May cause cancer by inhalation.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF ON SKIN: Wash with plenty of soap and water
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 235-852-4
RUO – Research Use Only