missing translation for 'onlineSavingsMsg'
Learn More

Chromium(II) chloride, 99.9%, (trace metal basis), Thermo Scientific Chemicals

Chemical Identifiers

CAS 10049-05-5
Molecular Formula Cl2Cr
Molecular Weight (g/mol) 122.9
MDL Number MFCD00010947
Synonym Chromous chloride
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10251433
View Documents Promotion Details
Thermo Scientific Acros
315190010
1 g Glass bottle N/A
N/A
10761161
View Documents Promotion Details
Thermo Scientific Acros
315190050
5 g Glass bottle N/A
N/A
10187413
View Documents Promotion Details
Thermo Scientific Acros
315190250
25 g Glass bottle N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis, Reduction reagent
Specifications

Chemical Identifiers

10049-05-5
122.9
Chromous chloride
Cl2Cr
MFCD00010947

Specifications

824.0°C
Gray-Green to White
99.9%
CrCl2
01,149; 02,76; 03,60; 04,144; 07,73; 09,511; 12,136; 13,84; 14,94; 16,93; 17,84
Chromous chloride
122.9
99.9%
Chromium(II) chloride
10049-05-5
Powder
Cl2Cr
MFCD00010947
15, 2248
Solubility in water: may decompose. Other solubilities: insoluble in alcohol and ether
122.9
Trace Metal Basis
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Harmful if swallowed.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 233-163-3

Product Identifier
  • Chromium(II) chloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only