missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) acetate hydroxide, Cr 24%, Thermo Scientific Chemicals

Product Code. 11331849
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Occasionally to dehalogenate organic compounds. used as an oxygen scrubber, polymer industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39430-51-8
Molecular Formula Cr3(OH)2(OOCCH3)7
MDL Number MFCD00058912

Specifications

Color Blue
Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water. Insoluble in acetone. Soluble in hot water
Formula Weight 603.31
Odor Odorless
Chemical Name or Material Chromium(III) acetate hydroxide, Cr 24%

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'