Learn More
Chromium(III) chloride, anhydrous, 99.9% (metals basis), Thermo Scientific Chemicals
Chemical Identifiers
CAS | 10025-73-7 |
---|---|
Molecular Formula | CrCl3 |
MDL Number | MFCD00010948 |
Synonym | chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0f |
Description
The vapor-phase co-reductions with other metal halides by hydrogen results in finely divided intermetallics with applications as structural materials or compounds with useful thermoelectric, magnetic, and oxidation-resistance properties. It is used as a catalyst, for organic synthesis, for manufacturing dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
10025-73-7 | |
CrCl3 | |
MFCD00010948 | |
chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0f |
Specifications
∽1150°C | |
Violet | |
99.9% (Metals basis) | |
MFCD00010948 | |
Soluble in alcohol,water. | |
Odorless | |
2.87 |
10025-73-7 | |
Flakes | |
CrCl3 | |
chromium iii chloride, chromium 3+ trichloride, anhydrous chromium iii chloride, basic chromic chloride, chromium chloride, basic, chromium trichl, chromium iii ; chloride, acmc-1ai6u, ksc492k0f | |
158.35 | |
Hygroscopic | |
Chromium(III) chloride |
Safety and Handling
P234-P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P390-P501c
H290-H302-H317
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: III
missing translation for 'einecsNumber' : 233-038-3
missing translation for 'rtecsNumber' : GB5425000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only