missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(III) Chloride Hexahydrate, Extra Pure, SLR, Fisher Chemical™
49.80 € - 134.00 €
Chemical Identifiers
CAS | 10060-12-5 |
---|---|
Molecular Formula | Cl3CrH12O6 |
Molecular Weight (g/mol) | 266.44 |
MDL Number | MFCD00149660 |
InChI Key | LJAOOBNHPFKCDR-UHFFFAOYSA-K |
IUPAC Name | chromium(3+) hexahydrate trichloride |
SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] |
Description
Chemical Identifiers
10060-12-5 | |
266.44 | |
LJAOOBNHPFKCDR-UHFFFAOYSA-K | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] |
Cl3CrH12O6 | |
MFCD00149660 | |
chromium(3+) hexahydrate trichloride |
Specifications
83°C | |
Green | |
Solid | |
MFCD00149660 | |
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cr+3] | |
266.44 | |
Glass Jar |
10060-12-5 | |
2 to 3 | |
Cl3CrH12O6 | |
LJAOOBNHPFKCDR-UHFFFAOYSA-K | |
chromium(3+) hexahydrate trichloride | |
266.44g/mol |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Chromium(III) chloride hexahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 C
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.