missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride tetrahydrofuran complex (1:3), 98%, Thermo Scientific Chemicals

Product Code. 10750945
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10170-68-0
Molecular Formula C12H24Cl3CrO3
Molecular Weight (g/mol) 374.67
MDL Number MFCD00013222
InChI Key CYOMBOLDXZUMBU-UHFFFAOYSA-K
Synonym chromium iii chloride tetrahydrofuran complex 1:3, chromium iii chloride tetrahydrofuran adduct, crcl3 thf 3, crcl3 3thf, chromium iii chloride tetrahydrofuran complex, chromium chloride; tris tetrahydrofuran, chromium iii chloride complex 1:3 tetrahydrofuran
PubChem CID 11100829
IUPAC Name oxolane;trichlorochromium
SMILES C1CCOC1.C1CCOC1.C1CCOC1.Cl[Cr](Cl)Cl

Specifications

Melting Point 143.0°C
Color Purple
Physical Form Crystalline Powder
Quantity 1 g
Assay Percent Range 98%
Formula Weight 374.67
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Chromium(III) chloride tetrahydrofuran complex (1:3)
compliance-icons compliance-icons
Product Identifier
  • Chromium(III) chloride tetrahydrofuran complex (1:3)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P381-Eliminate all ignition sources if safe to do so.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.