missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) chloride tetrahydrofuran complex (1:3), 98%, Thermo Scientific Chemicals

Chemical Identifiers

CAS 10170-68-0
Molecular Formula C12H24Cl3CrO3
Molecular Weight (g/mol) 374.67
MDL Number MFCD00013222
InChI Key CYOMBOLDXZUMBU-UHFFFAOYSA-K
Synonym chromium iii chloride tetrahydrofuran complex 1:3, chromium iii chloride tetrahydrofuran adduct, crcl3 thf 3, crcl3 3thf, chromium iii chloride tetrahydrofuran complex, chromium chloride; tris tetrahydrofuran, chromium iii chloride complex 1:3 tetrahydrofuran
PubChem CID 11100829
IUPAC Name oxolane;trichlorochromium
SMILES C1CCOC1.C1CCOC1.C1CCOC1.Cl[Cr](Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10750945
View Documents Promotion Details
Thermo Scientific Acros
389850010
1 g Glass bottle N/A
N/A
10702984
View Documents Promotion Details
Thermo Scientific Acros
389850100
10 g Glass bottle N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

10170-68-0
374.67
CYOMBOLDXZUMBU-UHFFFAOYSA-K
11100829
C1CCOC1.C1CCOC1.C1CCOC1.Cl[Cr](Cl)Cl
C12H24Cl3CrO3
MFCD00013222
chromium iii chloride tetrahydrofuran complex 1:3, chromium iii chloride tetrahydrofuran adduct, crcl3 thf 3, crcl3 3thf, chromium iii chloride tetrahydrofuran complex, chromium chloride; tris tetrahydrofuran, chromium iii chloride complex 1:3 tetrahydrofuran
oxolane;trichlorochromium

Specifications

143.0°C
Purple
98%
MFCD00013222
CYOMBOLDXZUMBU-UHFFFAOYSA-K
oxolane;trichlorochromium
11100829
98%
Chromium(III) chloride tetrahydrofuran complex (1:3)
10170-68-0
Crystalline Powder
C12H24Cl3CrO3
chromium iii chloride tetrahydrofuran complex 1:3, chromium iii chloride tetrahydrofuran adduct, crcl3 thf 3, crcl3 3thf, chromium iii chloride tetrahydrofuran complex, chromium chloride; tris tetrahydrofuran, chromium iii chloride complex 1:3 tetrahydrofuran
C1CCOC1.C1CCOC1.C1CCOC1.Cl[Cr](Cl)Cl
374.67
374.67
Authentic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable solid.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
May form explosive peroxides.

GHS P Statement
Eliminate all ignition sources if safe to do so.
Take precautionary measures against static discharge.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protec

GHS Signal Word: Warning

Product Identifier
  • Chromium(III) chloride tetrahydrofuran complex (1:3)

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H228-Flammable solid.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P381-Eliminate all ignition sources if safe to do so.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only