Learn More
Chromium(III) oxide, 98+%, Thermo Scientific Chemicals
46.90 € - 118.00 €
Chemical Identifiers
CAS | 1308-38-9 |
---|---|
Molecular Formula | Cr2O3 |
Molecular Weight (g/mol) | 151.99 |
MDL Number | MFCD00010949 |
InChI Key | UOUJSJZBMCDAEU-UHFFFAOYSA-N |
Synonym | chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide |
PubChem CID | 517277 |
ChEBI | CHEBI:48242 |
SMILES | [O--].[O--].[O--].[Cr+3].[Cr+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11391629
|
Thermo Scientific Alfa Aesar
012286.36 |
500 g |
46.90 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301639
|
Thermo Scientific Alfa Aesar
012286.A3 |
2 kg |
118.00 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is employed in metallurgical processing and has applications in chrome plating, anodizing aluminum etc… It is also used as a pigment.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1308-38-9 | |
151.99 | |
UOUJSJZBMCDAEU-UHFFFAOYSA-N | |
517277 | |
[O--].[O--].[O--].[Cr+3].[Cr+3] |
Cr2O3 | |
MFCD00010949 | |
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide | |
CHEBI:48242 |
Specifications
3000°C | |
1308-38-9 | |
Powder | |
Cr2O3 | |
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide | |
UOUJSJZBMCDAEU-UHFFFAOYSA-N | |
oxo(oxochromiooxy)chromium | |
517277 | |
151.99 | |
2.551 | |
Chromium(III) oxide |
2435°C | |
Green | |
>98% | |
MFCD00010949 | |
Insoluble in water,alcohol,acetone and acids | |
[O--].[O--].[O--].[Cr+3].[Cr+3] | |
151.99 | |
CHEBI:48242 | |
Odorless | |
5.2 g/cm3at 20°C |
Safety and Handling
Keep container in a well ventilated place. Avoid contact with skin. Wear suitable gloves.
H302-H332-H317 : Harmful if swallowed. Harmful if inhaled. May cause an allergic skin reaction.
missing translation for 'einecsNumber' : 215-160-9
missing translation for 'rtecsNumber' : GB6475000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only