missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) oxide, Puratronic™, 99.97% (metals basis), Thermo Scientific Chemicals

62.01 € - 1934.63 €

Chemical Identifiers

CAS 1308-38-9
Molecular Formula Cr2O3
Molecular Weight (g/mol) 151.99
MDL Number MFCD00010949
InChI Key UOUJSJZBMCDAEU-UHFFFAOYSA-N
Synonym chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
PubChem CID 517277
ChEBI CHEBI:48242
SMILES [O--].[O--].[O--].[Cr+3].[Cr+3]
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11340939
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010688.06
5 g
62.01 €
5g
Please to purchase this item. Need a web account? Register with us today!
11350939
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010688.14
25 g
194.97 €
25g
Please to purchase this item. Need a web account? Register with us today!
11360939
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010688.22
100 g
551.75 €
100g
Please to purchase this item. Need a web account? Register with us today!
11330939
View Documents Promotion Details
Thermo Scientific Alfa Aesar
010688.36
500 g
1934.63 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Also known as green compound for buffing and polishing, it can be used as abrasives. Apart from being used as a precursor to the magnetic pigment viz., chromium dioxide, it is used in inks, glasses, and pigments (viridian). They are employed as plating agents and surface treating agents. It is a catalyst in the Phillips process of manufacture of high density polythene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1308-38-9
151.99
UOUJSJZBMCDAEU-UHFFFAOYSA-N
517277
[O--].[O--].[O--].[Cr+3].[Cr+3]
Cr2O3
MFCD00010949
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
CHEBI:48242

Specifications

3000°C
1308-38-9
Powder
MFCD00010949
chromium iii oxide, chromia, dichromium trioxide, chromium sesquioxide, chrome green, green chromium oxide, chromium 3+ oxide, green cinnabar, chrome ochre, chrome oxide
UOUJSJZBMCDAEU-UHFFFAOYSA-N
dichromium(3+) trioxidandiide
517277
151.99
Puratronic™
2.551
(metals basis)
2435°C
Green
Cr2O3
14,2234
Insoluble in water,alcohol,acetone,alkalies and acids.
[O--].[O--].[O--].[Cr+3].[Cr+3]
151.99
CHEBI:48242
99.97%
Odorless
5.22 g/mL
Chromium(III) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H332-H317
Harmful if swallowed.
Harmful if inhaled.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

missing translation for 'einecsNumber' : 215-160-9

missing translation for 'rtecsNumber' : GB6475000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only