missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium(III) Potassium Sulfate Dodecahydrate, 98.0 to 102.0%, ACS Reagent, Honeywell Fluka™
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Melting Point | 89°C |
Color | Violet |
Quantity | 100 g |
Linear Formula | CrK(SO4)2 · 12H2O |
UN Number | NONH for all modes of transport |
Formula Weight | 499.4g/mol |
Percent Purity | 98.0 to 102.0% |
Grade | ACS Reagent |
Packaging | Poly Bottle |
Density | 1.8320g/cm3 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'