missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) Potassium Sulfate Dodecahydrate, 98.0 to 102.0%, ACS Reagent, Honeywell Fluka™

Product Code. 15612610
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7788-99-0
Molecular Formula CrH24KO20S2
Molecular Weight (g/mol) 499.386
MDL Number MFCD00149917
InChI Key ZFVHBEKVAITXHW-UHFFFAOYSA-J
Synonym chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
PubChem CID 24596
IUPAC Name potassium;chromium(3+);disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]

Specifications

Melting Point 89°C
Color Violet
Quantity 500 g
Linear Formula CrK(SO4)2 · 12H2O
UN Number NONH for all modes of transport
Formula Weight 499.4g/mol
Percent Purity 98.0 to 102.0%
Grade ACS Reagent
Packaging Poly Bottle
Density 1.8320g/cm3
Chemical Name or Material Chromium(Iii) Potassium Sulfate Dodecahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'