Learn More
Chromium(III) potassium sulfate dodecahydrate, 99.9% (metals basis), Thermo Scientific Chemicals
77.90 € - 1080.00 €
Chemical Identifiers
CAS | 7788-99-0 |
---|---|
Molecular Formula | CrH24KO20S2 |
Molecular Weight (g/mol) | 499.386 |
MDL Number | MFCD00149917 |
InChI Key | ZFVHBEKVAITXHW-UHFFFAOYSA-J |
Synonym | chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p |
PubChem CID | 24596 |
IUPAC Name | potassium;chromium(3+);disulfate;dodecahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11355178
|
Thermo Scientific Alfa Aesar
044083.14 |
25 g |
77.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11375178
|
Thermo Scientific Alfa Aesar
044083.22 |
100 g |
272.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11365178
|
Thermo Scientific Alfa Aesar
044083.36 |
500 g | |||||||
Description
It is used for tanning of leather and as a photographic film hardener. It is also used in textile dye, photography, ceramics, and the preparation of chromium standard solutions, as a catalyst, mordant, ceramics, tannage and developer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7788-99-0 | |
499.386 | |
ZFVHBEKVAITXHW-UHFFFAOYSA-J | |
24596 | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3] |
CrH24KO20S2 | |
MFCD00149917 | |
chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p | |
potassium;chromium(3+);disulfate;dodecahydrate |
Specifications
89°C | |
Violet | |
99.9% (Metal basis) | |
MFCD00149917 | |
Soluble in water 24g/100 mL (15°C). | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3] | |
499.386 | |
499.39 (283.21 Anhydrous) | |
Violet crystals | |
Chromium(III) potassium sulfate dodecahydrate |
7788-99-0 | |
Crystalline | |
CrH24KO20S2 | |
chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p | |
ZFVHBEKVAITXHW-UHFFFAOYSA-J | |
potassium;chromium(3+);disulfate;dodecahydrate | |
24596 | |
Odorless | |
1.83 g/cm3 |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 233-401-6
missing translation for 'rtecsNumber' : GB6850000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only