missing translation for 'onlineSavingsMsg'
Learn More

Chromium(III) sulfate hydrate, Reagent Grade, Thermo Scientific Chemicals

88.54 € - 346.73 €

Chemical Identifiers

CAS 15244-38-9
Molecular Formula Cr2O12S3
Molecular Weight (g/mol) 392.16
MDL Number MFCD00016039
InChI Key GRWVQDDAKZFPFI-UHFFFAOYSA-H
IUPAC Name dichromium(3+) trisulfate
SMILES [Cr+3].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11307628
View Documents
Thermo Scientific Alfa Aesar
033303.22
100 g
88.54 €
100g
Please to purchase this item. Need a web account? Register with us today!
11397618
View Documents
Thermo Scientific Alfa Aesar
033303.36
500 g
346.73 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Chromium(III) sulfate hydrate is widely used in making chrome tanned leather, paints, inks and ceramics. It is also used in preparation of chromium metal, dye, in printing and dyeing industry. It is involved in the preparation of chrome catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

15244-38-9
392.16
GRWVQDDAKZFPFI-UHFFFAOYSA-H
[Cr+3].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
Cr2O12S3
MFCD00016039
dichromium(3+) trisulfate

Specifications

15244-38-9
Crystals
MFCD00016039
GRWVQDDAKZFPFI-UHFFFAOYSA-H
dichromium(3+) trisulfate
392.17 (Anhydrous)
Odorless
Chromium(III) sulfate hydrate
Purple
Cr2O12S3
Soluble in water and alcohol. Insoluble in acids.
[Cr+3].[Cr+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
392.16
Reagent
Hygroscopic
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P333+P313-P362-P501c

H302+H332-H315-H317-H319

missing translation for 'einecsNumber' : 233-253-2

missing translation for 'rtecsNumber' : GB7200000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • Chromium(III) sulphate hydrate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Chromium(III) sulphate hydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.