missing translation for 'onlineSavingsMsg'
Learn More

Chromium pieces, 2-3mm (0.08-0.12in), 99.995% (metals basis), Thermo Scientific Chemicals

118.59 € - 420.09 €

Chemical Identifiers

CAS 7440-47-3
Molecular Formula Cr
Molecular Weight (g/mol) 52.00
MDL Number MFCD00010944
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
Synonym chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
PubChem CID 23976
ChEBI CHEBI:28073
IUPAC Name chromium
SMILES [Cr]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11338837
View Documents Promotion Details
Thermo Scientific Alfa Aesar
038494.18
50 g
118.59 €
50g
Please to purchase this item. Need a web account? Register with us today!
11348837
View Documents Promotion Details
Thermo Scientific Alfa Aesar
038494.30
250 g
420.09 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-47-3
52.00
VYZAMTAEIAYCRO-UHFFFAOYSA-N
23976
chromium
Cr
MFCD00010944
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
CHEBI:28073
[Cr]

Specifications

7440-47-3
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chromium
23976
99.995%
Broken Plate
Chromium pieces, 2 to 3mm (0.08 to 0.12 in.) thick
Cr
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
[Cr]
52.00
CHEBI:28073
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-157-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only