missing translation for 'onlineSavingsMsg'
Learn More

Chromium pieces, irregular, 99% (metals basis), Thermo Scientific Chemicals

77.08 € - 218.09 €

Chemical Identifiers

CAS 7440-47-3
Molecular Formula Cr
Molecular Weight (g/mol) 52.00
MDL Number MFCD00010944
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
Synonym chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
PubChem CID 23976
ChEBI CHEBI:28073
IUPAC Name chromium
SMILES [Cr]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11361208
SDS View Documents
Thermo Scientific Alfa Aesar
010150.22
100 g
77.08 €
100g
Please to purchase this item. Need a web account? Register with us today!
11351208
SDS View Documents
Thermo Scientific Alfa Aesar
010150.36
500 g
218.09 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-47-3
52.00
VYZAMTAEIAYCRO-UHFFFAOYSA-N
23976
chromium
Cr
MFCD00010944
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
CHEBI:28073
[Cr]

Specifications

Gray
Gray pieces
Cr
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
[Cr]
52.00
CHEBI:28073
0 hPa
Chromium
7440-47-3
99% (Metals basis)
MFCD00010944
VYZAMTAEIAYCRO-UHFFFAOYSA-N
chromium
23976
Odorless
Broken Plate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-157-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.