missing translation for 'onlineSavingsMsg'
Learn More

Chromium potassium sulfate dodecahydrate, 98+%, Thermo Scientific Chemicals

30.75 € - 265.32 €

Chemical Identifiers

CAS 7788-99-0
Molecular Formula CrKO8S2·12H2O
Molecular Weight (g/mol) 499.39
MDL Number MFCD00149917
InChI Key ZFVHBEKVAITXHW-UHFFFAOYSA-J
Synonym chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
PubChem CID 24596
IUPAC Name potassium;chromium(3+);disulfate;dodecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10012890
SDS View Documents
Thermo Scientific Acros
222520050
5 g Glass bottle
30.75 €
5g
Please to purchase this item. Need a web account? Register with us today!
10657372
SDS View Documents
Thermo Scientific Acros
222521000
100 g Plastic bottle
35.48 €
100g
Please to purchase this item. Need a web account? Register with us today!
10616222
SDS View Documents
Thermo Scientific Acros
222525000
500 g Plastic bottle
60.10 €
500g
Please to purchase this item. Need a web account? Register with us today!
10472835
SDS View Documents
Thermo Scientific Acros
222520025
2.5 kg Plastic bottle
265.32 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7788-99-0
499.39
ZFVHBEKVAITXHW-UHFFFAOYSA-J
24596
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]
CrKO8S2·12H2O
MFCD00149917
chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
potassium;chromium(3+);disulfate;dodecahydrate

Specifications

89.0°C
Purple
98.0 to 102% (Iodometry)
CrK(SO4)2·12H2O
15, 2233
Solubility in water: 24 g/100 mL (15°C).
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[K+].[Cr+3]
499.39
499.39
1.83
Chromium potassium sulfate dodecahydrate, p.a.
7788-99-0
Crystals
CrKO8S2·12H2O
MFCD00149917
chromium potassium sulfate dodecahydrate, chrome alum dodecahydrate, potassium chromium alum dodecahydrate, unii-h54d055wx6, ccris 8929, chromic potassium sulfate dodecahydrate, chromium iii potassium sulfate 1:1:2 , dodecahydrate, sulfuric acid, chromium 3+ potassium salt 2:1:1 , dodecahydrate, acmc-20alga, ksc492e4p
ZFVHBEKVAITXHW-UHFFFAOYSA-J
potassium;chromium(3+);disulfate;dodecahydrate
24596
≥98%
1.8300g/mL
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medi

GHS Signal Word: Warning

missing translation for 'rtecsNumber' : GB6850000

Product Identifier
  • Chromium potassium sulfate dodecahydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.