missing translation for 'onlineSavingsMsg'
Learn More

Chromium powder, -200 mesh, 99.94% (metals basis), Thermo Scientific Chemicals

Product Code. 11371329
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chromium, Cr, CAS Number-7440-47-3, chrome, chromium, elemental, ccris 159, chrome french, compounds, chrom, chrom german, cromo, metal, chromium, metal, 500g, Gray powder, 99.95% (Metals basis), CHEBI:28073, DOT Class: 4.1, Packaging Group: III, 231-157-5, H228

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-47-3
Molecular Formula Cr
Molecular Weight (g/mol) 52.00
MDL Number MFCD00010944
InChI Key VYZAMTAEIAYCRO-UHFFFAOYSA-N
Synonym chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159
PubChem CID 23976
ChEBI CHEBI:28073
IUPAC Name chromium
SMILES [Cr]

Specifications

Color Gray
Appearance Gray powder
Density 7.19g/cm3
Assay Percent Range 99.4% (Metals basis)
Quantity 100 g
Mesh Size -200 Mesh
Odor Odorless
Form Powder
Chemical Name or Material Chromium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'