missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromium powder, -60 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
134.00 € - 494.00 €
Chemical Identifiers
CAS | 7440-47-3 |
---|---|
Molecular Formula | Cr |
Molecular Weight (g/mol) | 52.00 |
MDL Number | MFCD00010944 |
InChI Key | VYZAMTAEIAYCRO-UHFFFAOYSA-N |
Synonym | chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 |
PubChem CID | 23976 |
ChEBI | CHEBI:28073 |
IUPAC Name | chromium |
SMILES | [Cr] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11475320
|
Thermo Scientific Alfa Aesar
044556.22 |
100 g |
134.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11485320
|
Thermo Scientific Alfa Aesar
044556.36 |
500 g |
494.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-47-3 | |
52.00 | |
VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
23976 | |
chromium |
Cr | |
MFCD00010944 | |
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
CHEBI:28073 | |
[Cr] |
Specifications
Gray | |
Gray powder | |
Cr | |
chrom, chrome, cromo, metal, chrome french, compounds, chrom german, chromium, elemental, chromium, metal, ccris 159 | |
[Cr] | |
52.00 | |
CHEBI:28073 | |
Odorless | |
Chromium |
7440-47-3 | |
99.99% (Metals basis) | |
MFCD00010944 | |
VYZAMTAEIAYCRO-UHFFFAOYSA-N | |
chromium | |
23976 | |
-60 Mesh | |
Powder |
Safety and Handling
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: III
missing translation for 'einecsNumber' : 231-157-5
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only