missing translation for 'onlineSavingsMsg'
Learn More

Chromium selenide, Black powder, 99.5% (Metals basis), Thermo Scientific Chemicals

Product Code. 11382208
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used in semiconductor, chemical vapor deposition (CVD) and physical vapor deposition (PVD) display and optical applications. It is used for coating solar energy or fuel cells and flip-chip applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12053-13-3
Molecular Formula CrSe
Molecular Weight (g/mol) 130.967
MDL Number MFCD00049885
InChI Key UVZCKRKEVWSRGT-UHFFFAOYSA-N
Synonym chromium selenide, chromium ii selenide
PubChem CID 82886
IUPAC Name selanylidenechromium
SMILES [Cr]=[Se]

Specifications

Color Black
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.5% (Metals basis)
Formula Weight 130.96
Odor Odorless
Density 6.74g/cm3
Chemical Name or Material Chromium selenide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'