Learn More
Chromium silicide, 99+% (metals basis), Thermo Scientific Chemicals
65.30 € - 235.00 €
Chemical Identifiers
CAS | 12018-09-6 |
---|---|
Molecular Formula | CrSi2 |
Molecular Weight (g/mol) | 108.166 |
MDL Number | MFCD00168070 |
InChI Key | CHXGWONBPAADHP-UHFFFAOYSA-N |
Synonym | chromsilicid, chromium silicide, bis $l^ 3 |
PubChem CID | 45051936 |
IUPAC Name | bis($l^{3}-silanylidyne)chromium |
SMILES | [Si]#[Cr]#[Si] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11336528
|
Thermo Scientific Alfa Aesar
012151.18 |
50 g |
65.30 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326528
|
Thermo Scientific Alfa Aesar
012151.30 |
250 g |
235.00 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chromium silicide is applied in production of ceramic materials. It can also produce an oxidation-resistant surface in a low pressure plasma spraying.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12018-09-6 | |
108.166 | |
CHXGWONBPAADHP-UHFFFAOYSA-N | |
45051936 | |
[Si]#[Cr]#[Si] |
CrSi2 | |
MFCD00168070 | |
chromsilicid, chromium silicide, bis $l^ 3 | |
bis($l^{3}-silanylidyne)chromium |
Specifications
1490°C | |
Gray | |
>95% | |
MFCD00168070 | |
Insoluble in water. | |
[Si]#[Cr]#[Si] | |
108.166 | |
108.17 | |
Moisture sensitive | |
Chromium silicide |
12018-09-6 | |
Powder | |
CrSi2 | |
chromsilicid, chromium silicide, bis $l^ 3 | |
CHXGWONBPAADHP-UHFFFAOYSA-N | |
bis($l^{3}-silanylidyne)chromium | |
45051936 | |
Odorless | |
Gray powder |
Sicherheit und Handhabung
P261-P264b-P271-P272-P280-P285-P302+P352-P304+P340-P305+P351+P338-P333+P313-P342+P311-P362-P501c
H315-H317-H319-H334-H335
missing translation for 'dotInformation' : Hazard Class: 9; Packaging Group: III
missing translation for 'einecsNumber' : 234-633-0
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only