missing translation for 'onlineSavingsMsg'
Learn More

Chromium silicon monoxide, 99.8+% (metals basis), Thermo Scientific™

Product Code. 11337828
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

For vacuum deposition

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56488-64-3
MDL Number MFCD02091702

Specifications

Physical Form Sintered Pieces
Quantity 25 g
Solubility Information Insoluble in water.
Percent Purity ≥99.8%
Assay (metals basis)
Chemical Name or Material Chromium silicon monoxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'