Learn More
Chromium silicon monoxide, 99.85+% (metals basis), Thermo Scientific™
For vacuum deposition
90.50 € - 327.00 €
Chemical Identifiers
CAS | 70320-14-8 |
---|---|
MDL Number | MFCD02091702 |
Synonym | chromium silicon monoxide, d-arg1,d-trp5 inverted exclamation mark currency7 inverted exclamation mark currency9,leu11-substance p |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
70320-14-8 | |
MFCD02091702 | |
chromium silicon monoxide, d-arg1,d-trp5 inverted exclamation mark currency7 inverted exclamation mark currency9,leu11-substance p |
Specifications
70320-14-8 | |
MFCD02091702 | |
Insoluble in water. | |
(metals basis) |
Sintered Pieces | |
chromium silicon monoxide, d-arg1,d-trp5 inverted exclamation mark currency7 inverted exclamation mark currency9,leu11-substance p | |
≥99.85% | |
Chromium silicon monoxide |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.