missing translation for 'onlineSavingsMsg'
Learn More

Chromium silicon monoxide, 99.9% (metals basis), Thermo Scientific™

Product Code. 11313827
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37264-20-3
Molecular Formula CrOSi
Molecular Weight (g/mol) 96.08
MDL Number MFCD02091702
InChI Key ASAMIKIYIFIKFS-UHFFFAOYSA-N
Synonym chromium silicon monoxide, d-arg1,d-trp5 inverted exclamation mark currency7 inverted exclamation mark currency9,leu11-substance p
PubChem CID 57464841
IUPAC Name chromium;oxoniumylidynesilanide
SMILES [O+]#[Si-].[Cr]

Specifications

Quantity 10 g
Solubility Information It is insoluble in water.
Percent Purity 99.9%
Assay (metals basis)
Chemical Name or Material Chromium silicon monoxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.