missing translation for 'onlineSavingsMsg'
Learn More

Chromosulfuric acid, Thermo Scientific Chemicals

70.45 €

Chemical Identifiers

CAS 65272-71-1
Molecular Formula Cr2K2O7·H2O4S
Molecular Weight (g/mol) 200.237
MDL Number MFCD00010952
InChI Key IFMFTIBWWVBKOC-UHFFFAOYSA-L
Synonym absorbency linearity solution #2 24.28 mg/l inverted exclamation marka in hclo4, 25 ml
PubChem CID 131664149
IUPAC Name dipotassium;chromium;dihydroxide;dihydrate
SMILES O.O.[OH-].[OH-].[K+].[K+].[Cr]
View More Specs

Products 1
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10722961
View Documents
Thermo Scientific Acros
295510010
1 L Glass bottle
70.45 €
1 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
Specifications

Chemical Identifiers

65272-71-1
200.237
IFMFTIBWWVBKOC-UHFFFAOYSA-L
131664149
O.O.[OH-].[OH-].[K+].[K+].[Cr]
Cr2K2O7·H2O4S
MFCD00010952
absorbency linearity solution #2 24.28 mg/l inverted exclamation marka in hclo4, 25 ml
dipotassium;chromium;dihydroxide;dihydrate

Specifications

Brown to Brown-Red
330.0°C
1.84
Liquid
absorbency linearity solution #2 24.28 mg/l inverted exclamation marka in hclo4, 25 ml
IFMFTIBWWVBKOC-UHFFFAOYSA-L
dipotassium;chromium;dihydroxide;dihydrate
131664149
65272-71-1
1.8400g/mL
Cr2K2O7·H2O4S
MFCD00010952
Solubility in water: soluble in water (caution ! development of heat)
O.O.[OH-].[OH-].[K+].[K+].[Cr]
200.237
Chromosulfuric acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful to aquatic life with long lasting effects.
May cause an allergic skin reaction.
May cause damage to organs through prolonged or repeated exposure.
May cause c

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Danger

Product Identifier
  • n-Butylmagnesium chloride, 20 wt% solution in THF/Toluene

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Carcinogenicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Tetrahydrofuran, Toluene, Butylmagnesiumchloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.