Learn More
Proteintech ChromoTek Halo-Trap Magnetic Agarose Kit
The ChromoTek Halo-Trap Magnetic Agarose Kit consists of an anti-Halo-tag Nanobody (VHH), which is covalently bound to magnetic agarose beads.
Brand:  Proteintech otmak-20
Product Code. 17353413
  • 497.00 € / 20 reactions
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Halo-Trap Magnetic Agarose Kit is used to immunoprecipitate Halo-tagged fusion proteins from cell extracts of various organisms like mammals, plants, bacteria, yeast, insects etc. in the absence or presence of a covalently bound ligand
  • The interaction between Halo-Trap and the Halo-tagged fusion protein is reversible
Specifications
Specifications
20 Reactions