Learn More
Proteintech ChromoTek mNeonGreen-Trap Magnetic Agarose Kit
Kit including 20 reactions of mNeonGreen-Trap Magnetic Agarose and lysis, dilution, wash, and elution buffers.
Brand:  Proteintech ntmak-20
Product Code. 17323403
  • 497.00 € / 20 reactions
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The ChromoTek mNeonGreen-Trap Magnetic Agarose is an affinity resin for IP of mNeonGreen-fusion proteins
  • Consists of a mNeonGreen Nanobody/ VHH coupled to magnetic agarose beads
Specifications
Specifications
20 Reactions
20 Reactions