missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Proteintech ChromoTek Nano-CaptureLigand™ Human IgE, Recombinant VHH, Biotinylated
Nano-CaptureLigand for human IgE: single domain antibody/VHH [CTK0111], biotinylated.
Brand: Proteintech shuEB-1-100
This item is not returnable.
View return policy
Description
- The ChromoTek Nano-CaptureLigand™ human IgE, VHH, biotinylated is used for the site-directed and specific immobilization of human IgE in biosensor and ELISA assays
- It captures non-biotinylated human IgE antibodies to streptavidin/avidin
- Nano-CaptureLigand human IgE, VHH, biotinylated comprises a monoclonal biotinylated VHH/ Nanobody
Specifications
100 μL | |
Biotin | |
Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F |
Fc-fragment of Human IgG1, IgG2, IgG3, and IgG4, Fc-fragment of Rabbit IgG | |
Monoclonal | |
BLI, SPR, ELISA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction