Learn More
Proteintech ChromoTek Nano-CaptureLigand™ Human IgG/Rabbit IgG, Fc-specific VHH, Biotinylated
Nano-CaptureLigand for human IgG and rabbit IgG: Fc-specific single domain antibody/VHH [CTK0101], biotinylated.
Brand:  Proteintech shurbGB-1-10
 View more versions of this product
Product Code. 17373463
  • 113.00 € / 10µL
Explore more special offers
Description
Description
  • The Nano-CaptureLigand™ human IgG captures and immobilizes the Fc-fragment of human IgG antibodies to avidin/streptavidin coated surfaces used in BLI, SPR, and ELISA assays
  • Consists of a biotinylated Nanobody that specifically binds to human IgG antibodies with high affinity and in a site-directed manner
Specifications
Specifications
10 μL
Biotin
Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F
Human IgE
Monoclonal
BLI, SPR, ELISA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Proteintech ChromoTek Nano-CaptureLigand™ Human IgG/Rabbit IgG, Fc-specific VHH, Biotinylated > 10 μL