Learn More
Proteintech ChromoTek Nano-CaptureLigand™ Mouse IgG2a, Fc-specific VHH, Biotinylated
Nano-CaptureLigand for mouse IgG2a: Fc-specific single domain antibody/VHH [CTK0108], biotinylated.
Brand:  Proteintech smsG2aB-1-100
 View more versions of this product
Product Code. 17303493
  • 426.00 € / 100µL
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The Nano-CaptureLigand mouse IgG2a captures and immobilizes mouse IgG2a antibodies and Fc-fragments to avidin/streptavidin coated surfaces used in BLI, SPR, and ELISA assays
  • Consists of a biotinylated Nanobody that specifically binds to the mouse IgG2a subclass with high affinity and in a site-directed manner
Specifications
Specifications
100 μL
Biotin
Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F
Fc-fragment of Mouse IgG2a
Monoclonal
BLI, SPR, ELISA