Learn More
Proteintech ChromoTek Nano-CaptureLigand™ Mouse IgG2b, Fc-specific VHH, Biotinylated
Nano-CaptureLigand for mouse IgG2b: Fc-specific single domain antibody/VHH [CTK0106], biotinylated.
Brand:  Proteintech smsG2bB-1-10
 View more versions of this product
Product Code. 17313493
  • 114.00 € / 10µL
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The Nano-CaptureLigand mouse IgG2b captures and immobilizes mouse IgG2b antibodies and Fc-fragments to avidin/streptavidin coated surfaces used in BLI, SPR, and ELISA assays
  • Consists of a biotinylated Nanobody that specifically binds to mouse IgG2b antibodies with high affinity and in a site-directed manner
Specifications
Specifications
10 μL
Biotin
Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F
Fc-fragment of Mouse IgG2b
Monoclonal
BLI, SPR, ELISA