missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek Nano-CaptureLigand™ Mouse IgG2b, Fc-specific VHH, Biotinylated

Nano-CaptureLigand for mouse IgG2b: Fc-specific single domain antibody/VHH [CTK0106], biotinylated.

Brand:  Proteintech SMSG2BB-1-100

283.34 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 17323493

  • 425.00 € / 100µL
In Stock
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

  • The Nano-CaptureLigand mouse IgG2b captures and immobilizes mouse IgG2b antibodies and Fc-fragments to avidin/streptavidin coated surfaces used in BLI, SPR, and ELISA assays
  • Consists of a biotinylated Nanobody that specifically binds to mouse IgG2b antibodies with high affinity and in a site-directed manner
Specifications

Specifications

100 μL
Biotin
Upon receipt store at +4°C/+40°F. Optional: Aliquot upon arrival and store at -20°C/-4°F
Fc-fragment of Mouse IgG2b
Monoclonal
BLI, SPR, ELISA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.