Learn More
Proteintech ChromoTek TurboGFP-Trap Magnetic Agarose Kit
Kit including 20 reactions of TurboGFP-Trap Magnetic Agarose and lysis, dilution, wash, and elution buffers.
Brand:  Proteintech tbtmak-20
Product Code. 17353523
  • 497.00 € / 20 reactions
Estimated Shipment: 06-05-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • TurboGFP-Trap Magnetic Agarose is an affinity reagent for immunoprecipitation (IP) of TurboGFP fusion proteins
  • Consists of an anti-TurboGFP VHH/ Nanobody that is coupled to magnetic agarose beads
Specifications
Specifications
20 Reactions
20 Reactions