missing translation for 'onlineSavingsMsg'
Learn More

Proteintech ChromoTek V5-Trap™ Magnetic Agarose

The ChromoTek V5-Trap™ Magnetic Agarose is an affinity resin for IP of V5-tagged proteins.

238.00 € - 5899.00 €

Specifications

Content And Storage Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year.
For Use With (Application) IP, CoIP, ChIP, RIP
Target V5-tag sequence GKPIPNPLLGLDST at the N-, C-terminal or internal tagging.
Label or Dye Magnetic Agarose
Species Alpaca
View More Specs

Products 5
Product Code Brand Sufficient For Price Quantity & Availability  
Product Code Brand Sufficient For Price Quantity & Availability  
17353543
View Documents
Proteintech
v5tma-10
10 Reactions
238.00 €
10 reactions
In Stock
to see stock.
Add to basket
 
17363543
View Documents
Proteintech
v5tma-100
100 Reactions
1798.00 €
100 reactions
In Stock
to see stock.
Add to basket
 
17373543
View Documents
Proteintech
v5tma-20
20 Reactions
397.00 €
20 reactions
In Stock
to see stock.
Add to basket
 
17383543
View Documents
Proteintech
v5tma-200
200 Reactions
3237.00 €
200 reactions
In Stock
to see stock.
Add to basket
 
17393543
View Documents
Proteintech
v5tma-400
400 Reactions
5899.00 €
400 reactions
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
 
Description

Description

  • Consists of a anti-V5 Nanobody/VHH coupled to magnetic agarose beads
Specifications

Specifications

Shipped at ambient temperature. Upon receipt store at 4°C. Stable for one year.
V5-tag sequence GKPIPNPLLGLDST at the N-, C-terminal or internal tagging.
Alpaca
IP, CoIP, ChIP, RIP
Magnetic Agarose
Videos
SDS
Documents

Documents

Product Certifications