Learn More
Chromplete™ Methanol, HPLC/GC, meets ACS/USP analytical requirements, Thermo Scientific™
The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Methanol is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry.
21.50 € - 41.40 €
Chemical Identifiers
CAS | 67-56-1 |
---|---|
Molecular Formula | CH4O |
Molecular Weight (g/mol) | 32.04 |
MDL Number | MFCD00004595 |
InChI Key | OKKJLVBELUTLKV-UHFFFAOYSA-N |
Synonym | methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit |
PubChem CID | 887 |
ChEBI | CHEBI:17790 |
IUPAC Name | methanol |
SMILES | CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15990711
|
Fisher Scientific Chemical Div
T001021000 |
1 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15900721
|
Fisher Scientific Chemical Div
T001022500 |
2.5 L |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15910721
|
Fisher Scientific Chemical Div
T001024000 |
4 L |
41.40 €
4 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The Chromplete product line from Thermo Scientific includes reagents for common analyses and separations with the consistency and precision you expect. Chromplete Methanol is suitable for HPLC, GC, SFC, TLC, ACS, and Spectrophotometry.
Chromplete Methanol meets the analytical requirements of USP and is suitable for use in:
- HPLC coupled with UV, VIS, PDA, ECD, FL, and RI detectors
- GC- FID, GC- ECD, GC-TCD
- Pesticide Analysis by GC or HPLC
- UV Spectrophotometry
- Preparative chromatography
- Supercritical Fluid and Thin Layer chromatography
- Reactions/Analyses requiring ACS compliance
- Extraction/Concentration Analyses
Chemical Identifiers
67-56-1 | |
32.04 | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
887 | |
methanol |
CH4O | |
MFCD00004595 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CHEBI:17790 | |
CO |
Specifications
-98°C | |
67-56-1 | |
≤ 5 APHA | |
148.5°F | |
Liquid | |
CH4O | |
≤0.0002 meq/g | |
1230 | |
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit | |
CO | |
32.04 | |
CHEBI:17790 | |
99.9% | |
Minimum, GC-FID |
≤1.00, ≤ 0.23, ≤ 0.11, ≤ 0.01 | |
0.999 | |
0.775 – 0.780 | |
Amber Glass | |
≤ 1.0 ppm | |
≤0.0003 meq/g | |
MFCD00004595 | |
≤0.07% | |
OKKJLVBELUTLKV-UHFFFAOYSA-N | |
methanol | |
887 | |
32.04 | |
Chromplete | |
Methanol |
Safety and Handling
Highly flammable liquid and vapor, Causes damage to organs, Toxic if swallowed, in contact with skin or if inhaled
Flammable liquids (category 2), Specific target organ toxicity after single exposure (category 1), Acute toxicity (category 3)
missing translation for 'shelfLife' : 1825 days
missing translation for 'einecsNumber' : 200-659-6
missing translation for 'rtecsNumber' : PC1400000
missing translation for 'storageNote' : In sealed containers
Recommended Storage : Room temperature flammable storage
- Chromplete Methanol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
- MIXTURE LIST-Contains : methanol
RUO – Research Use Only