Learn More
Chromyl chloride, 99.99% (metals basis), Thermo Scientific Chemicals
Description
Highly spin-polarized chromium dioxide (CrO2) thin films were deposited on 100 TiO2 substrates by chemical vapor deposition using chromyl chloride as a precursor. Chromyl chloride (Cr02C12) reacts with cyclohexane solvent at 75 OC to give a dark precipitate along with chlorocyclohexane and a small amount of cyclohexene (in 10.0 and ca. 0.3% yields based on chromium). Chromyl chloride, CrO2C12, oxidizes cyclooctane, isobutane, and toluene under mild conditions (25-60 C). The reactions give chlorinated products (chlorocyclooctane, tert- butyl chloride, and benzyl chloride) and a dark chromium-containing precipitate. Used for Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins. Chromyl chloride in carbon tetrachloride solution reacts in the cold with cyclohexene, cyclopentene, and 1-hexene to give the various isomeric chlorohydrins as the major products.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Organic oxidations and chlorination, chromium coordination complexes, catalyst for polymerization of olefins

Specifications
Specifications
Boiling Point | 117°C |
Melting Point | -96.5°C |
Physical Form | Liquid |
Quantity | 50 g |
UN Number | UN1758 |
Merck Index | 14,2248 |
Solubility Information | Soluble in carbon tetrachloride,carbon disulfide,benzene,nitrobenzene,chloroform,and POCL3. Insoluble in water(Reacts). |
Formula Weight | 154.9 |
Percent Purity | 99.99% |
Odor | Pungent, makes eyes water |
Show More |
Product Suggestions
Customers who viewed this item also viewed
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Z24K031 | Certificate of Analysis | 26/12/2024 | 040517.09,040517.18 |
R29K005 | Certificate of Analysis | 11/06/2024 | 040517.09,040517.18 |
W06J066 | Certificate of Analysis | 21/09/2023 | 040517.09,040517.18 |
T25J035 | Certificate of Analysis | 07/08/2023 | 040517.09,040517.18 |
Y15I052 | Certificate of Analysis | 29/11/2022 | 040517.09,040517.18 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.