Learn More
Tocris Bioscience™ CID 1375606
GPR27 agonist
Brand:  Tocris Bioscience™ 6544/25
 View more versions of this product
Product Code. 18750878
  • 290.00 € / 25mg
Explore more special offers
Description
Description
CID 1375606 is a GPR27 agonist. Promotes recruitment of β-arrestin 2 to chimeric GPR27V2 and wild-type GPR27 in a firefly luciferase complementation assay (pEC50 values are 6.34 and 5.1, respectively).
Specifications
Specifications
2,4-Dichloro-N-(phenylcarbamoyl)phenylbenzamide
25 mg
98%
313493-80-0
C20H14Cl2N2O2