missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Selective protein kinase D (PKD) inhibitor (IC50 values are 0.182, 0.280, 0.227, >10, 15.3, 20.3, 40.5 and >50 μM at PKD1, PKD2, PKD3, PKC, CAK, PLK1, CAMKIIα and Akt respectively). Blocks PKD-mediated protein transport and inhibits prostate cancer cell proliferation, migration and invasion in vitro.

Specifications
Specifications
CAS | 521937-07-5 |
Molecular Formula | C12H11NO3 |
MDL Number | MFCD03828155 |
Quantity | 10 mg |
Synonym | cid, 7-hydroxy-2,3,4,5-tetrahydro-1h-benzofuro 2,3-c azepin-1-one, 7-hydroxy-2,3,4,5-tetrahydro-1h-1 benzofuro 2,3-c azepin-1-one, 2,3,4,5-tetrahydro-7-hydroxy-1h-benzofuro 2,3-c azepin-1-one, 7-hydroxy-2,3,4,5-tetrahydro-1 benzofuro 2,3-c azepin-1-one, 3-hydroxy-5,6,7,8-tetrahydro-10-oxa-8-aza-benzo a azulen-9-one, 4-hydroxy-8-oxa-11-azatricyclo 7.5.0.0 2 ,? tetradeca-1 9 ,2,4,6-tetraen-10-one, d0mc3k, regid_for_cid_755673, cid hplc , powder |
InChI Key | AACFPJSJOWQNBN-UHFFFAOYSA-N |
SMILES | OC1=CC=C2OC3=C(CCCNC3=O)C2=C1 |
IUPAC Name | 4-hydroxy-8-oxa-11-azatricyclo[7.5.0.0²,⁷]tetradeca-1(9),2,4,6-tetraen-10-one |
Molecular Weight (g/mol) | 217.22 |
PubChem CID | 755673 |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CID 755673, Tocris Bioscience™ >
Spot an opportunity for improvement?Share a Content Correction