missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Cidofovir dihydrate

231.15 € - 686.42 €

Specifications

Format Powder
Molecular Weight (g/mol) 315.22
Product Type Cidofovir dihydrate
Source synthetic source
Content And Storage Freezer -20°C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16641852
View Documents
Thermo Scientific Chemicals
463120010
1 g
686.42 €
1g
Please to purchase this item. Need a web account? Register with us today!
16651852
View Documents
Thermo Scientific Chemicals
463122500
250 mg
231.15 €
250mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Powder
Cidofovir dihydrate
Freezer -20°C
≥97.5 % by HPLC
315.22
synthetic source
normal conditions
White to almost white
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 2 years

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.