Learn More
Thermo Scientific™ Cimarec™ 10M Controllers
Adjust speed from 100 to 1000rpm with this external control unit required for use with the Mobil 10 and Mobil 25 stirrers.
Brand:  Thermo Scientific™ 50119120
 View more versions of this product
Product Code. 12066319
  • 331.65 € / Each
Estimated Shipment: 20-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Adjust speed from 100 to 1000rpm with the Thermo Scientific™ Cimarec™ 10M Controller, an external control unit that is required for use with the Mobil 10 and Mobil 25 stirrers.
  • Automatic start-up for safe stirrer acceleration
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mobil 10 and Mobil 25 Stirrer
Stirrer Controller
230 V
100 to 1000 rpm
230 V
100 RPM to 1000 RPM