missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cimarec™ Biosystem 40B Controller

Enable synchronized operation of two stirrers with Thermo Scientific™ Cimarec™ Biosystem 40B Controller.

Brand:  Thermo Scientific™ 50118917

 View more versions of this product

Product Code. 12425771

  • 1587.90 € / Each
Estimated Shipment: 31-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Enable synchronized operation of two stirrers with Thermo Scientific™ Cimarec™ Biosystem 40B Controller.

  • High visibility display
  • Adjustable stirring and pause times with rotation sense inversion
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Biomodul 40B Controller 230V Denmark
Stirrer Controller (Dual)
230 V
Biomodul 40B Controller
230 V Denmark
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.