Learn More
Thermo Scientific™ Cimarec™ i Benchtop Distributors (Link and Sync)
Ensure identical operating conditions for multiple stirrer installations by using these benchtop distributors, for use with Mini, Micro, Compact and Maxi series stirrers.
235.00 € - 345.00 €
Specifications
For Use With (Equipment) | Mini, Micro, Compact, and Maxi Series Stirrer |
---|---|
Type | Benchtop Distributor |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10489391
|
Thermo Scientific™
50091720 |
4-Place Distributor |
235.00 €
Each |
In Stock
Log in to see stock. |
|||||
10247081
|
Thermo Scientific™
50091721 |
8-Place Distributor |
345.00 €
Each |
In Stock
Log in to see stock. |
|||||
Description
Ensure identical operating conditions for multiple stirrer installations by using Thermo Scientific™ Cimarec™ i Benchtop Distributors (Link and Sync), for use with Mini, Micro, Compact and Maxi series stirrers.
- Simultaneous control of 2, 4, 6, or 8 Mini, Micro or Compact series stirrers using a single Telemodul 20 C or Telemodul 40 C external controller
- Simultaneous control of 2 or 4 Maxi Series stirrers using a single Telemodul 20 C or Telemodul 40 C external controller
- Even-powered distribution for synchronized stirring
Specifications
Mini, Micro, Compact, and Maxi Series Stirrer | |
Benchtop Distributor |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.