missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl alcohol, 98% trans, Thermo Scientific Chemicals

Product Code. 10746361
Click to view available options
Quantity:
5 g
100 g
250 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-54-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00002921
InChI Key OOCCDEMITAIZTP-QPJJXVBHSA-N
Synonym cinnamyl alcohol, cinnamic alcohol, 3-phenyl-2-propen-1-ol, 3-phenylprop-2-en-1-ol, zimtalcohol, styryl carbinol, e-3-phenylprop-2-en-1-ol, 3-phenylallyl alcohol, trans-cinnamyl alcohol, styryl alcohol
PubChem CID 5315892
ChEBI CHEBI:33227
IUPAC Name (E)-3-phenylprop-2-en-1-ol
SMILES C1=CC=C(C=C1)C=CCO

Specifications

Melting Point 31.0°C to 35.0°C
Color White
Density 1.0400g/mL
Boiling Point 258.0°C
Flash Point 126°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula C6H5CH=CHCH2OH
Quantity 5 g
Beilstein 06, 570
Merck Index 15, 2303
Specific Gravity 1.04
Solubility Information Solubility in water: 1.8g/L (20°C). Other solubilities: soluble in glycerol, methanol, clearly soluble in 3 volumes 50% alcohol, freely soluble in alcohol, ether, and other, common organic solvents
Formula Weight 134.18
Percent Purity 98% trans
Physical Form Fused Low Melting Crystalline Solid
Chemical Name or Material Cinnamyl alcohol, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Cinnamyl alcohol
Signal Word
  • Warning
Hazard Category
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'