missing translation for 'onlineSavingsMsg'
Learn More

Steris™ CIP 220™

Formulated to meet the unique cleaning demands in industries requiring an effective, validatable detergent. Steris Corporation CIP 220™ is a proprietary blend of hydroxyacetic acid, an advanced surfactant and other performance-enhancing ingredients that provide multiple cleaning mechanisms.

Brand:  Steris™ 1D2208


Product Code. 15252680

This item is not returnable. View return policy

Description

Description

CIP 220 Acid-Based Process & Research Cleaner is specially formulated to meet the unique cleaning demands found in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries. This phosphate free low foaming detergent is ideal for use in CIP, COP and manual applications.

  • Flexibility—Effective cleaning against a wide variety of pharmaceutical process residues
  • Easy to Validate—Facilitates cleaning validation with a comprehensive support documentation package
  • Cost Savings—Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
  • Environmentally Friendly—Phosphate free and manufactured with biodegradable surfactants to support harmonization of cleaning processes
  • Reliability—Global availability and manufactured for the pharmaceutical industry in a cGMP-compliant, FDA-registered and ISO 9001-certified facility
Specifications

Specifications

Liquid
Colorless to Slightly Yellow
4 Pk.
3.8 L
2.8
10–30% hydroxyacetic acid
Completely soluble in water
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • CIP 220

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

Supplemental information
  • MIXTURE LIST-Contains : Hydroxyacetic acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.