missing translation for 'onlineSavingsMsg'
Learn More
Learn More
cis-1,2-Cyclohexanediol, 99%, Thermo Scientific Chemicals
43.72 € - 144.72 €
Chemical Identifiers
CAS | 1792-81-0 |
---|---|
Molecular Formula | C6H12O2 |
Molecular Weight (g/mol) | 116.16 |
MDL Number | MFCD00064944 |
InChI Key | PFURGBBHAOXLIO-OLQVQODUSA-N |
Synonym | cis-1,2-cyclohexanediol, cis-cyclohexane-1,2-diol, grandidentol, cis-1,2-dihydroxycyclohexane, 1r,2s-cyclohexane-1,2-diol, 1,2-cyclohexanediol, cis, 1,2-cyclohexanediol, cis-8ci 9ci, 1,2-cyclohexanediol, 1r,2s-rel, cis-1,2-cyclohexandiol, 1alpha,2alpha-cyclohexanediol |
PubChem CID | 92903 |
ChEBI | CHEBI:32329 |
IUPAC Name | (1R,2S)-cyclohexane-1,2-diol |
SMILES | C1CCC(C(C1)O)O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10003450
|
Thermo Scientific Acros
300970010 |
1 g | Glass bottle |
43.72 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10412855
|
Thermo Scientific Acros
300970050 |
5 g | Glass bottle |
144.72 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1792-81-0 | |
116.16 | |
PFURGBBHAOXLIO-OLQVQODUSA-N | |
92903 | |
(1R,2S)-cyclohexane-1,2-diol |
C6H12O2 | |
MFCD00064944 | |
cis-1,2-cyclohexanediol, cis-cyclohexane-1,2-diol, grandidentol, cis-1,2-dihydroxycyclohexane, 1r,2s-cyclohexane-1,2-diol, 1,2-cyclohexanediol, cis, 1,2-cyclohexanediol, cis-8ci 9ci, 1,2-cyclohexanediol, 1r,2s-rel, cis-1,2-cyclohexandiol, 1alpha,2alpha-cyclohexanediol | |
CHEBI:32329 | |
C1CCC(C(C1)O)O |
Specifications
1792-81-0 | |
White to Beige | |
98.5% min. (GC) | |
C6H10(OH)2 | |
cis-1,2-cyclohexanediol, cis-cyclohexane-1,2-diol, grandidentol, cis-1,2-dihydroxycyclohexane, 1r,2s-cyclohexane-1,2-diol, 1,2-cyclohexanediol, cis, 1,2-cyclohexanediol, cis-8ci 9ci, 1,2-cyclohexanediol, 1r,2s-rel, cis-1,2-cyclohexandiol, 1alpha,2alpha-cyclohexanediol | |
PFURGBBHAOXLIO-OLQVQODUSA-N | |
(1R,2S)-cyclohexane-1,2-diol | |
92903 | |
116.16 | |
Crystalline Flakes or Powder |
98°C to 101°C | |
Authentic | |
C6H12O2 | |
MFCD00064944 | |
Solubility in water: slightly soluble in water | |
C1CCC(C(C1)O)O | |
116.16 | |
CHEBI:32329 | |
99% | |
cis-1, 2-Cyclohexanediol |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title