missing translation for 'onlineSavingsMsg'
Learn More

cis-1,2-Dichloroethylene, 97%, stabilized, Thermo Scientific Chemicals

Product Code. 10684762
Click to view available options
Quantity:
2.5 mL
10 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 156-59-2
Molecular Formula C2H2Cl2
Molecular Weight (g/mol) 96.94
MDL Number MFCD00000925
InChI Key KFUSEUYYWQURPO-UPHRSURJSA-N
Synonym cis-1,2-dichloroethene, z-1,2-dichloroethene, cis-1,2-dichloroethylene, z-1,2-dichloroethylene, cis-dichloroethylene, acetalyne dichloride, acetylene dichloride, cis, ethene, 1,2-dichloro-, z, cis-acetylene dichloride, unii-fyo9g15jyd
PubChem CID 643833
ChEBI CHEBI:28805
IUPAC Name (Z)-1,2-dichloroethene
SMILES C(=CCl)Cl

Specifications

Melting Point -80.0°C
Color Colorless
Density 1.2800g/mL
Boiling Point 60.0°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClCH=CHCl
Refractive Index 1.4475 to 1.4495
Quantity 10 mL
Beilstein 01,188
Fieser 13,293
Merck Index 15,87
Specific Gravity 1.28
Stabilizer 0.003% MEHQ
Formula Weight 96.94
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material cis-1, 2-Dichloroethylene, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.