missing translation for 'onlineSavingsMsg'
Learn More

cis-2,6-Dimethylpiperidine, 97+%, Thermo Scientific™

Product Code. 10326700
Click to view available options
Quantity:
5 g
100 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-17-6
Molecular Formula C7H15N
Molecular Weight (g/mol) 113.2
MDL Number MFCD00005986
InChI Key SDGKUVSVPIIUCF-KNVOCYPGSA-N
Synonym cis-2,6-dimethylpiperidine, 2r,6s-2,6-dimethylpiperidine, cis-hexahydro-2,6-lutidine, cis-2,6-lupetidine, piperidin, 2e,6e-dimethyl, cis-2,6-dimethyl-piperidine, cis-lupetidine, cis-nanophine, prestwick0_000666, prestwick1_000666
PubChem CID 641768
IUPAC Name (2R,6S)-2,6-dimethylpiperidine
SMILES CC1CCCC(N1)C

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8400g/mL
Boiling Point 127°C to 128°C
Flash Point 11°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4384 to 1.4404
Quantity 100 g
UN Number 2733
Beilstein 20, 108
Fieser 02,156
Specific Gravity 0.84
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most organic solvents
Formula Weight 113.2
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material cis-2, 6-Dimethylpiperidine, 99%, min. 95% cis-isomer
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • cis-2,6-Dimethylpiperidine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.